Novus Biologicals
Manufacturer Code:NBP252566
Catalog # NBP252566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C8D3 |
Immunogen | Synthesized peptide of human SK2 (AA: 36-52). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.91 SK 2 sphingosine kinase 2 sphingosine kinase type 2 isoform SPK 2; SK 2; Sphingosine kinase 2; sphingosine kinase type 2
Gene Aliases: SK 2; SK-2; SK2; SPHK2; SPK 2; SPK-2
UniProt ID: (Human) Q9NRA0
Entrez Gene ID: (Human) 56848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.