Novus Biologicals
Manufacturer Code:NBP274332AF647
Catalog # N274332A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A6 |
Immunogen | Full length recombinant protein of human Sphingosine Kinase 1/SPHK1 (NP_068807) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acetyltransferase SPHK1; EC 2.7.1.91 sphingosine kinase 1 SPHKSK 1 SPK SPK 1; SK 1; Sphingosine kinase 1; SPK 1
Gene Aliases: SK1; SPHK; SPHK1; SPK
UniProt ID: (Human) Q9NYA1
Entrez Gene ID: (Human) 8877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.