R&D Systems
Manufacturer Code:MAB55361100
Catalog # MAB55361100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 647705 |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line Sf 21-derived recombinant human Sphingosine Kinase 1/SPHK1 Asp2-Leu398 Accession No. Q9NYA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: Acetyltransferase SPHK1; EC 2.7.1.91 SK1 sphingosine kinase 1 SPHKSK 1 SPK SPK 1 SPK1; SK 1; Sphingosine kinase 1; SPK 1
Gene Aliases: SK1; SPHK; SPHK1; SPK
UniProt ID: (Human) Q9NYA1
Entrez Gene ID: (Human) 8877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.