Novus Biologicals
Manufacturer Code:NBP160004
Catalog # NBP160004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SGMS2(sphingomyelin synthase 2) The peptide sequence was selected from the N terminal of SGMS2. Peptide sequence KFPLEWWKTGIAFIYAVFNLVLTTVMITVVHERVPPKELSPPLPDKFFDY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.8.27 phosphatidylcholine:ceramide cholinephosphotransferase 2 SM synthase sphingomyelin synthase 2SMS2MGC26963; Phosphatidylcholine:ceramide cholinephosphotransferase 2; SM synthase; Sphingomyelin synthase 2
Gene Aliases: SGMS2; SMS2
UniProt ID: (Human) Q8NHU3
Entrez Gene ID: (Human) 166929
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.