Proteintech
Manufacturer Code:198581AP150UL
Catalog # 50-173-2578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Spermidine synthase Fusion Protein Ag13936 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAPT SPDSY Spermidine synthase SPS1 SRM SRML1; Putrescine aminopropyltransferase; SPDSY; Spermidine synthase; spermidine synthase-1
Gene Aliases: AA407669; PAPT; SpdST; SPDSY; SPS1; SRM; SRML1
UniProt ID: (Human) P19623, (Mouse) Q64674
Entrez Gene ID: (Human) 6723, (Mouse) 20810, (Rat) 84596
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.