Novus Biologicals
Manufacturer Code:NBP254440200UG
Catalog # NB254440200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPTBN2/1583 |
Immunogen | Recombinant human SPTBN2 fragment (aa356-475) (exact sequence is proprietary) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-III spectrin; Beta-III spectrin glutamate transporter EAAT4-associated protein 41 GTRAP41 KIAA0302 SCA5 spectrin beta chain brain 2 spectrin beta non-erythrocytic 2 Spectrin non-erythroid beta chain 2 spinocerebellar ataxia 5; glutamate transporter EAAT4-associated protein 41; spectrin beta chain, brain 2; Spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; Spinocerebellar ataxia 5 protein
Gene Aliases: GTRAP41; KIAA0302; SCA5; SCAR14; SPTBN2
UniProt ID: (Human) O15020
Entrez Gene ID: (Human) 6712
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.