Invitrogen
Manufacturer Code:711584
Catalog # 71-158-4
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Clone | 11HCLC |
Immunogen | Peptides corresponding to Human SPTBN2 (aa 1334-1351 1733-1750) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-III spectrin; beta-III spectrin beta-spectrin 3 glutamate transporter EAAT4-associated protein 41 GTRAP41 KIAA0302 mKIAA0302 SCA5 SCAR14 Spectrin Beta spectrin beta 3 spectrin beta chain brain 2 spectrin beta chain non-erythrocytic 2 spectrin beta III sigma 2 spectrin beta non-erythrocytic 2 spectrin beta non-erythrocytic 2 spectrin non-erythroid beta chain 2 spinocerebellar ataxia 5 protein Spnb3 SPTBN2; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; spectrin beta 3; spectrin beta chain, brain 2; Spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; Spinocerebellar ataxia 5 protein
Gene Aliases: GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spnb3; SPTBN2
UniProt ID: (Human) O15020
Entrez Gene ID: (Human) 6712, (Mouse) 20743
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.