Novus Biologicals
Manufacturer Code:NB11058346SS
Catalog # NB11058346T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The epitope recognized by this antibody maps to a C-terminal region between residue 2340 and 2390 of Spectrin beta 3 using the numbering given in entry NP_008877.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-III spectrin; Beta-III spectrin glutamate transporter EAAT4-associated protein 41 GTRAP41 KIAA0302 SCA5 spectrin beta chain brain 2 spectrin beta non-erythrocytic 2 Spectrin non-erythroid beta chain 2 spinocerebellar ataxia 5; glutamate transporter EAAT4-associated protein 41; spectrin beta chain, brain 2; Spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; Spinocerebellar ataxia 5 protein
Gene Aliases: GTRAP41; KIAA0302; SCA5; SCAR14; SPTBN2
UniProt ID: (Human) O15020
Entrez Gene ID: (Human) 6712
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.