Novus Biologicals
Manufacturer Code:NB10040828
Catalog # NB10040828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 2100 and 2150 of human Spectrin Non-Erythroid Beta 1 using the numbering given in entry NP_003119.2 (GeneID 6711). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-fodrin; beta-fodrin beta-G spectrin Beta-II spectrin beta-spectrin 2 beta-spectrin II betaSpII ELF embryonic liver beta-fodrin Fodrin beta chain spectrin beta chain brain 1 spectrin beta non-erythrocytic 1 Spectrin non-erythroid beta chain 1 SPTB2; beta-G spectrin; Beta-II spectrin; beta-spectrin 2; beta-spectrin II; beta-spectrin non-erythrocytic 1; embryonic liver beta-fodrin; epididymis luminal protein 102; Fodrin beta chain; spectrin beta chain, brain 1; Spectrin beta chain, non-erythrocytic 1; spectrin, beta, non-erythrocytic 1; Spectrin, non-erythroid beta chain 1
Gene Aliases: betaSpII; ELF; HEL102; SPTB2; SPTBN1
UniProt ID: (Human) Q01082
Entrez Gene ID: (Human) 6711
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.