Novus Biologicals
Manufacturer Code:NBP250307IR
Catalog # NBP250307IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RG/26 |
Immunogen | Red cell membranes |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-I spectrin; beta-I spectrin EL3 erythrocyte HS2 HSPTB1 membrane cytoskeletal protein spectrin beta erythrocytic SPTB1; membrane cytoskeletal protein; Sp beta; spectrin beta chain, erythrocyte; Spectrin beta chain, erythrocytic; spectrin beta Tandil
Gene Aliases: EL3; HS2; HSPTB1; SPH2; SPTB; SPTB1
UniProt ID: (Human) P11277
Entrez Gene ID: (Human) 6710
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.