Novus Biologicals
Manufacturer Code:NBP250303FR
Catalog # NBP250303FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Sp 6C6 |
Immunogen | Recombinant Spastin |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADPSPSpastic paraplegia 4 protein FSP2EC 3.6.4.3 KIAA1083EC 3.6.1.3 spastic paraplegia 4 (autosomal dominant spastin) spastin SPG4; SPAST; spastic paraplegia 4 (autosomal dominant; spastin); Spastic paraplegia 4 protein; Spastin
Gene Aliases: ADPSP; FSP2; KIAA1083; SPAST; SPG4
UniProt ID: (Human) Q9UBP0
Entrez Gene ID: (Human) 6683
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.