Invitrogen
Manufacturer Code:14981182
Catalog # 50-112-9095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | Btjce |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ANOP3 MCOPS3 SRY-related HMG-box gene 2 transcription factor SOX-2 transcription factor SOX2 sex determining region Y box 2; sex determining region Y-box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2
Gene Aliases: ANOP3; lcc; MCOPS3; Sox-2; SOX2; ysb
UniProt ID: (Human) P48431, (Mouse) P48432
Entrez Gene ID: (Human) 6657, (Mouse) 20674
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.