Invitrogen
Manufacturer Code:PA572000
Catalog # PIPA572000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 169-196 amino acids from the C-terminal region of human SOSTDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cystine-knot containing secreted protein; Ectodermal BMP inhibitor; Ectodin; Sclerostin domain-containing protein 1; Sclerostin domain-containing protein 1 Ectodermal BMP inhibitor Ectodin Uterine sensitization-associated gene 1 protein USAG-1 SOSTDC1 USAG1; Sclerostin-like protein; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1
Gene Aliases: 0610006G05Rik; CDA019; DAND7; ECTODIN; SOSTDC1; Sostl; USAG-1; USAG1; Wise
UniProt ID: (Human) Q6X4U4, (Mouse) Q9CQN4
Entrez Gene ID: (Human) 25928, (Mouse) 66042
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.