Invitrogen
Manufacturer Code:PA570641
Catalog # PIPA570641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse Sostdc1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cystine-knot containing secreted protein; Ectodermal BMP inhibitor; Ectodin; Sclerostin domain-containing protein 1; Sclerostin-like protein; Sostdc1 Sostl Usag1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1; Wnt-signaling modulator
Gene Aliases: 0610006G05Rik; CDA019; DAND7; ECTODIN; SOSTDC1; Sostl; USAG-1; USAG1; Wise
UniProt ID: (Human) Q6X4U4, (Mouse) Q9CQN4, (Rat) Q642G2
Entrez Gene ID: (Human) 25928, (Dog) 100856691, (Horse) 100052877, (Rabbit) 100354321, (Bovine) 523184, (Mouse) 66042, (Rat) 266803, (Guinea pig) 100716997
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.