Invitrogen
Manufacturer Code:PA519481
Catalog # PIPA519481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 800 to the C-terminus of Human Sortilin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa NT receptor; Glycoprotein 110; Glycoprotein 95; Gp110; Gp95; Gp95 LDLCQ6 NT3 100 kDa NT receptor NTR3 glycoprotein 95 neurotensin receptor 3 sortilin; mNTR3; Neurotensin receptor 3; NT3; NTR3; Sortilin
Gene Aliases: 2900053A11Rik; AI852375; Gp95; LDLCQ6; NT3; NTR3; Nts3; Ntsr3; SORT1
UniProt ID: (Human) Q99523, (Mouse) Q9QXW6, (Rat) O54861
Entrez Gene ID: (Human) 6272, (Mouse) 20661, (Rat) 83576
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.