Invitrogen
Manufacturer Code:OSS00052W
Catalog # PIOSS00052W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the extracellular N-terminal part of rat Sortilin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein 110; Gp110; mNTR3; Neurotensin receptor 3; Neurotensin receptor 3 NTR3 Glycoprotein 110 Gp110 Nt3 Nts3; NTR3; Sortilin
Gene Aliases: 2900053A11Rik; AI852375; Nt3; Ntr3; Nts3; Ntsr3; Sort1
UniProt ID: (Mouse) Q9QXW6, (Rat) O54861
Entrez Gene ID: (Mouse) 20661, (Rat) 83576
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.