R&D Systems
Manufacturer Code:AF3154SP
Catalog # AF3154SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa NT receptor; 100 kDa NT receptor Glycoprotein 95 Gp95 Gp95LDLCQ6 Neurotensin receptor 3 NT3NTR3 Ntr3 SORT1 sortilin sortilin 1; Glycoprotein 95; Gp95; Neurotensin receptor 3; NT3; Sortilin
Gene Aliases: Gp95; LDLCQ6; NT3; NTR3; SORT1
UniProt ID: (Human) Q99523
Entrez Gene ID: (Human) 6272
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.