Proteintech
Manufacturer Code:123691AP150UL
Catalog # 50-172-7932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sortilin Fusion Protein Ag2997 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa NT receptor; 100 kDa NT receptor Glycoprotein 95 Gp95 Neurotensin receptor 3 NT3 NTR3 SORT1 Sortilin sortilin 1; Glycoprotein 110; Glycoprotein 95; Gp110; Gp95; mNTR3; Neurotensin receptor 3; NT3; NTR3; Sortilin
Gene Aliases: 2900053A11Rik; AI852375; Gp95; LDLCQ6; NT3; NTR3; Nts3; Ntsr3; SORT1
UniProt ID: (Human) Q99523, (Mouse) Q9QXW6, (Rat) O54861
Entrez Gene ID: (Human) 6272, (Mouse) 20661, (Rat) 83576
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.