Invitrogen
Manufacturer Code:PA564975
Catalog # PIPA564975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Sorcin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa protein; calcium binding protein amplified in mutlidrug-resistant cells; CP-22; CP-22 CP22 SCN V19 22 kDa protein H&lowbarRG167B05.1 calcium binding protein amplified in mutlidrug-resistant cells; CP22; H_RG167B05.1; Sorcin; V19
Gene Aliases: CP-22; CP22; SCN; SRI; V19
UniProt ID: (Human) P30626
Entrez Gene ID: (Human) 6717
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.