Proteintech
Manufacturer Code:158811AP150UL
Catalog # 50-173-0550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sorbitol dehydrogenase Fusion Protein Ag8651 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; L iditol 2 dehydrogenase SDH Sorbitol dehydrogenase SORD SORD1; L-iditol 2-dehydrogenase; Polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; XDH; Xylitol dehydrogenase
Gene Aliases: HEL-S-95n; Sdh-1; Sdh1; Sodh-1; SORD; SORD1
UniProt ID: (Human) Q00796, (Rat) P27867, (Mouse) Q64442
Entrez Gene ID: (Human) 6652, (Rat) 24788, (Mouse) 20322
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.