Novus Biologicals
Manufacturer Code:NBP274288PECY7
Catalog # N27428PECY7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9E7 |
Immunogen | Full length recombinant protein of human Sorbitol Dehydrogenase (NP_003095) produced in HEK293T cells. |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: (R,R)-butanediol dehydrogenase; EC 1.1.1 EC 1.1.1.14 L-iditol 2-dehydrogenase sorbitol dehydrogenase SORD1; epididymis secretory sperm binding protein Li 95n; L-iditol 2-dehydrogenase; Polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sorbitol dehydrogenase; XDH; Xylitol dehydrogenase
Gene Aliases: HEL-S-95n; SORD; SORD1
UniProt ID: (Human) Q00796
Entrez Gene ID: (Human) 6652
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.