Invitrogen
Manufacturer Code:PA545178
Catalog # PIPA545178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SORD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; L-iditol 2-dehydrogenase; Polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; Sorbitol dehydrogenase SORD1; XDH; Xylitol dehydrogenase
Gene Aliases: HEL-S-95n; SDH; Sdh-1; Sdh1; Sodh-1; SORD; SORD1; XDH
UniProt ID: (Bovine) Q58D31, (Human) Q00796, (Rat) P27867, (Mouse) Q64442
Entrez Gene ID: (Rabbit) 100355885, (Sheep) 101115280, (Dog) 487535, (Horse) 100070575, (Bovine) 508954, (Pig) 100158181, (Human) 6652, (Rat) 24788, (Mouse) 20322, (Guinea pig) 100712656
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.