Invitrogen
Manufacturer Code:PA518333
Catalog # PIPA518333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KIMLKCDPSDQNP) corresponding to the C-terminus amino acids of SORD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: L-iditol 2-dehydrogenase; Polyol dehydrogenase; SDH; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; sorbitol dehydrogenase SORD SORD1; XDH; Xylitol dehydrogenase
Gene Aliases: Sdh-1; Sdh1; Sodh-1; Sord
UniProt ID: (Mouse) Q64442
Entrez Gene ID: (Mouse) 20322
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.