Novus Biologicals
Manufacturer Code:NBP128885
Catalog # NBP128885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7D7B11 |
Immunogen | Purified recombinant fragment of SORL1(aa2159-2214) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C11orf32 chromosome 11 open reading frame 32 FLJ21930 FLJ39258 gp250 Low-density lipoprotein receptor relative with 11 ligand-binding repeats LR11LDLR relative with 11 ligand-binding repeats LRP9 mosaic protein LR11 SorLA SorLA-1SORLA sortilin-related receptor sortilin-related receptor L(DLR class) A repeats containing sortilin-related receptor L(DLR class) A repeats-containing Sorting protein-related receptor containing LDLR class A repeats; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mosaic protein LR11; SorLA; SorLA-1; Sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: C11orf32; gp250; LR11; LRP9; SORL1; SORLA; SorLA-1
UniProt ID: (Human) Q92673
Entrez Gene ID: (Human) 6653
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.