R&D Systems
Manufacturer Code:MAB5699SP
Catalog # MAB5699SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 525122 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human SorLA Ser82-Ser1550 Accession # Q92673 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C11orf32 chromosome 11 open reading frame 32 FLJ21930 FLJ39258 gp250 Low-density lipoprotein receptor relative with 11 ligand-binding repeats LR11 LR11LDLR relative with 11 ligand-binding repeats LRP9 mosaic protein LR11 SORL1 SorLA SorLA-1SORLA sortilin-related receptor sortilin-related receptor L(DLR class) A repeats containing sortilin-related receptor L(DLR class) A repeats-containing Sorting protein-related receptor containing LDLR class A repeats; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mosaic protein LR11; SorLA; SorLA-1; Sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: C11orf32; gp250; LR11; LRP9; SORL1; SORLA; SorLA-1
UniProt ID: (Human) Q92673
Entrez Gene ID: (Human) 6653
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.