R&D Systems
Manufacturer Code:AF4237SP
Catalog # AF4237SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse SorCS2 Ala118-Gly1078 Accession # Q9EPR5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1329VPS10 domain receptor protein sortilin-related VPS10 domain containing receptor 2 VPS10 domain-containing receptor SorCS2; SorCS2 104 kDa chain; SorCS2 122 kDa chain; SorCS2 18 kDa chain; sortilin-related VPS10 domain containing receptor 2; VPS10 domain receptor protein; VPS10 domain-containing receptor SorCS2
Gene Aliases: KIAA1329; SORCS2
UniProt ID: (Human) Q96PQ0
Entrez Gene ID: (Human) 57537
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.