Invitrogen
Manufacturer Code:PA552883
Catalog # PIPA552883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SorCS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ41758 FLJ43475 FLJ44957 hSorCS SORCS SORCS receptor 1 sorCS1 sortilin-related VPS10 domain containing receptor 1 VPS10 domain receptor protein SORCS 1 VPS10 domain receptor SorCS VPS10 domain-containing receptor SorCS1; hSorCS; sortilin-related VPS10 domain containing receptor 1; VPS10 domain receptor protein SORCS 1; VPS10 domain-containing receptor SorCS1
Gene Aliases: hSorCS; SORCS; SORCS1
UniProt ID: (Human) Q8WY21
Entrez Gene ID: (Human) 114815
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.