Invitrogen
Manufacturer Code:PA5113257
Catalog # PIPA5113257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human VPS10 domain-containing receptor SorCS1 protein (34-175AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ41758 FLJ43475 FLJ44957 hSorCS mSorCS SORCS SORCS receptor 1 SORCS1 sortilin related VPS10 domain containing receptor 1 sortilin-related VPS10 domain containing receptor 1 VPS10 domain receptor protein SORCS 1 VPS10 domain-containing receptor SorCS1; hSorCS; sortilin-related VPS10 domain containing receptor 1; VPS10 domain receptor protein SORCS 1; VPS10 domain-containing receptor SorCS1
Gene Aliases: hSorCS; SORCS; SORCS1
UniProt ID: (Human) Q8WY21
Entrez Gene ID: (Human) 114815
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.