Novus Biologicals
Manufacturer Code:NBP222126
Catalog # NBP222126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H4 |
Immunogen | Purified recombinant fragment of human Sonic Hedgehog (amino acids 26-170) expressed in E. coli. [UniProt# Q15465] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HHG-1; HHG1 HHG-1 HLP3 HPE3 MCOPCB5sonic hedgehog (Drosophila) homolog SMMCIsonic hedgehog homolog (Drosophila) sonic hedgehog sonic hedgehog homolog sonic hedgehog protein TPT TPTPS; SHH; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; ShhN; ShhNC; ShhNp; sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein N-product
Gene Aliases: HHG1; HLP3; HPE3; MCOPCB5; SHH; SMMCI; TPT; TPTPS
UniProt ID: (Human) Q15465
Entrez Gene ID: (Human) 6469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.