R&D Systems
Manufacturer Code:MAB4641500
Catalog # MAB4641500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 171018 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Sonic Hedgehog Cys25-Gly198 (Lys122Arg) Accession # Q62226 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: HHG-1; HHG1 HHG-1 HLP3 HPE3 MCOPCB5 MCOPCB5sonic hedgehog (Drosophila) homolog Shh ShhNC SMMCI SMMCIsonic hedgehog homolog (Drosophila) sonic hedgehog sonic hedgehog homolog sonic hedgehog protein TPT TPTPS; SHH; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; ShhN; ShhNC; ShhNp; sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein N-product
Gene Aliases: HHG1; HLP3; HPE3; MCOPCB5; SHH; SMMCI; TPT; TPTPS
UniProt ID: (Human) Q15465
Entrez Gene ID: (Human) 6469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.