Novus Biologicals
Manufacturer Code:NBP29523820UL
Catalog # NB024639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 314-388 of human SSTR4 (NP_001043.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled receptor; G-protein coupled receptor somatostatin receptor 4 somatostatin receptor type 4 SS4R SS-4-R SS4-R; Somatostatin receptor type 4; SS-4-R
Gene Aliases: SS-4-R; SS4-R; SS4R; SSTR4
UniProt ID: (Human) P31391
Entrez Gene ID: (Human) 6754
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.