R&D Systems
Manufacturer Code:IC4224U100UG
Catalog # IC4224U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 402038 |
Immunogen | NS0 mouse myeloma cell line transfected with Somatostatin R2/SSTR2 Met1-Ile369 Accession # P30874 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Smstr2 somatostatin receptor 2 somatostatin receptor type 2 SomatostatinR2 SRIF-1 SS2R SS-2-R SS2-R SST2 SSTR2; Somatostatin receptor type 2; SRIF-1; SS-2-R; SS2R
Gene Aliases: SSTR2
UniProt ID: (Human) P30874
Entrez Gene ID: (Human) 6752
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.