Novus Biologicals
Manufacturer Code:NBP227413
Catalog # NBP227413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was raised against Somatostatin Receptor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled receptor somatostatin receptor 1 somatostatin receptor type 1 SRIF-2 SS1R SS-1-R SS1-R SSR1; Somatostatin receptor type 1; SRIF-2; SS-1-R
Gene Aliases: SRIF-2; SS-1-R; SS1-R; SS1R; SSTR1
UniProt ID: (Human) P30872
Entrez Gene ID: (Human) 6751
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.