Novus Biologicals
Manufacturer Code:NBP29317120UL
Catalog # NB028230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human SST (NP_001039.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Growth hormone release-inhibiting factor; Growth hormone release-inhibiting factor SMST somatostatin somatostatin-14 somatostatin-28; Neuronostatin; NST; prepro-somatostatin; Somatostatin; Somatostatin-14; Somatostatin-28; SST-14
Gene Aliases: SMST; SST
UniProt ID: (Human) P61278
Entrez Gene ID: (Human) 6750
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.