Novus Biologicals
Manufacturer Code:NBP15725320UL
Catalog # NBP15725320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLA/LP(soluble liver antigen/liver pancreas antigen) The peptide sequence was selected from the N terminal of SLA/LP. Peptide sequence MDSNNFLGNCGVGEREGRVASALVARRHYRFIHGIGRSGDISAVQPKAAG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.9.1.2 EC 2.9.1.n1 Liver-pancreas antigen LPDKFZp434B1417 MGC161491 O-phosphoseryl-tRNA(Sec) selenium transferase Sec synthase Selenocysteine synthase Selenocysteinyl-tRNA(Sec) synthase Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase SepSecS Sep-tRNA:Sec-tRNA synthase SLA/LP SLA/LP autoantigen SLA-p35 SLAUGA suppressor tRNA-associated protein Soluble liver antigen soluble liver antigen/liver pancreas antigen tRNA(Ser/Sec)-associated antigenic protein TRNP48; Liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; Sec synthase; Selenocysteine synthase; Selenocysteinyl-tRNA(Sec) synthase; Sep-tRNA:Sec-tRNA synthase; SepSecS; SLA; SLA-p35; SLA/LP autoantigen; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; tRNA(Ser/Sec)-associated antigenic protein; UGA suppressor tRNA-associated protein
Gene Aliases: LP; PCH2D; SEPSECS; SLA; SLA/LP; TRNP48
UniProt ID: (Human) Q9HD40
Entrez Gene ID: (Human) 51091
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.