Novus Biologicals
Manufacturer Code:NB300540
Catalog # NB300540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | XVIF9-G10 |
Immunogen | Canine cardiac microsomes. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 3 polypeptide ATPase Na+/K+ transporting alpha 3 polypeptide dystonia 12 DYT12 EC 3.6.3 MGC13276 RDP Sodium pump subunit alpha-3 sodium/potassium-transporting ATPase alpha-3 chain sodium/potassium-transporting ATPase subunit alpha-3; ATPase, Na+/K+ transporting, alpha 3 polypeptide; Na(+)/K(+) ATPase alpha(III) subunit; Na(+)/K(+) ATPase alpha-3 subunit; Na+, K+ activated adenosine triphosphatase alpha subunit; Na+/K+ ATPase 3; Sodium pump subunit alpha-3; sodium-potassium ATPase catalytic subunit alpha-3; sodium-potassium-ATPase, alpha 3 polypeptide; sodium/potassium-transporting ATPase alpha-3 chain; Sodium/potassium-transporting ATPase subunit alpha-3
Gene Aliases: AHC2; ATP1A1; ATP1A3; CAPOS; DYT12; RDP
UniProt ID: (Human) P13637
Entrez Gene ID: (Human) 478
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.