Novus Biologicals
Manufacturer Code:NBP29466920UL
Catalog # NB023863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ATP1A2 (NP_000693.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-A(+) catalytic polypeptide ATPase Na+/K+ transporting alpha 2 (+) polypeptide ATPase Na+/K+ transporting alpha 2 polypeptide EC 3.6.3 EC 3.6.3.9 KIAA0778 MGC59864 MHP2 migraine hemiplegic 2 Na(+)/K(+) ATPase alpha-2 subunit Na+/K+ ATPase alpha-B polypeptide Sodium pump subunit alpha-2 sodium/potassium-transporting ATPase alpha-2 chain sodium/potassium-transporting ATPase subunit alpha-2 sodium-potassium ATPase; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase, Na+/K+ transporting, alpha 2 polypeptide; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; Sodium/potassium-transporting ATPase subunit alpha-2
Gene Aliases: ATP1A2; FHM2; KIAA0778; MHP2
UniProt ID: (Human) P50993
Entrez Gene ID: (Human) 477
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.