Novus Biologicals
Manufacturer Code:NBP29508920UL
Catalog # NB024345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 551-850 of human ATP1A (NP_000692.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase Na+/K+ transporting alpha 1 polypeptide EC 3.6.3 EC 3.6.3.9 K-ATPase alpha-1 subunit K-ATPase catalytic subunit alpha-A protein MGC3285 MGC51750 Na Na(+)/K(+) ATPase alpha-1 subunit Na K-ATPase alpha-A catalytic polypeptide Na+ K+ ATPase alpha subunit Na+/K+ ATPase 1 sodium pump 1 Sodium pump subunit alpha-1 sodium/potassium-transporting ATPase subunit alpha-1 sodium-potassium-ATPase alpha 1 polypeptide; ATPase, Na+/K+ transporting, alpha 1 polypeptide; Na(+)/K(+) ATPase alpha-1 subunit; Na+/K+ ATPase 1; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Sodium pump subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; Sodium/potassium-transporting ATPase subunit alpha-1
Gene Aliases: ATP1A1
UniProt ID: (Human) P05023
Entrez Gene ID: (Human) 476
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.