Invitrogen
Manufacturer Code:PA577723
Catalog # PIPA577723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide TEEQKKYYNAMKKLGSKK(C) corresponding to amino acid residues 1501-1518 of rat Nav 1.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative product; ataxia 3; brain III voltage-gated sodium channel; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CTC-264K15.6; HBSC II; hH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; mH1; Mu-1; Na+ channel; NaCh6; NaN; Neuroendocrine sodium channel; Peripheral nerve protein type 4; Peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; Peripheral sodium channel 1; PN1; PN3; PN4; PN5; RIIA sodium channel protein; Sensory neuron sodium channel; Sensory neuron sodium channel 2; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; SkM1; sodium channel 25; Sodium channel 6; sodium channel alpha-subunit; Sodium channel protein brain I subunit alpha; Sodium channel protein brain II subunit alpha; Sodium channel protein brain III subunit alpha; Sodium channel protein cardiac muscle subunit alpha; Sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 1 subunit alpha; Sodium channel protein type 10 subunit alpha; Sodium channel protein type 11 subunit alpha; Sodium channel protein type 2 subunit alpha; Sodium channel protein type 3 subunit alpha; Sodium channel protein type 4 subunit alpha; Sodium channel protein type 5 subunit alpha; Sodium channel protein type 8 subunit alpha; Sodium channel protein type 9 subunit alpha; Sodium channel protein type I subunit alpha; Sodium channel protein type II subunit alpha; Sodium channel protein type III subunit alpha; Sodium channel protein type IV subunit alpha; Sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; Sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; Sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type 4 alpha polypeptide; sodium channel voltage-gated type I alpha polypeptide; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type V alpha polypeptide; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type I alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage gated, type XI alpha subunit; sodium channel, voltage-gated, type 1, alpha polypeptide; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type 2, alpha 1 polypeptide; sodium channel, voltage-gated, type 2, alpha 1 subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type 4, alpha polypeptide; sodium channel, voltage-gated, type 4, alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type 8, alpha polypeptide; sodium channel, voltage-gated, type 8, alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type I, alpha polypeptide; sodium channel, voltage-gated, type I, alpha subunit; sodium channel, voltage-gated, type II, alpha 1; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha polypeptide 1; sodium channel, voltage-gated, type II, alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium channel, voltage-gated, type VIII, alpha polypeptide; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; SP-19; voltage gated sodium channel; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subtype II; Voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.1; Voltage-gated sodium channel subunit alpha Nav1.2; Voltage-gated sodium channel subunit alpha Nav1.3; Voltage-gated sodium channel subunit alpha Nav1.4; Voltage-gated sodium channel subunit alpha Nav1.5; Voltage-gated sodium channel subunit alpha Nav1.6; Voltage-gated sodium channel subunit alpha Nav1.7; Voltage-gated sodium channel subunit alpha Nav1.8; Voltage-gated sodium channel subunit alpha Nav1.9; voltage-gated sodium channel type II alpha subunit; voltage-gated sodium channel type VIII alpha protein
Gene Aliases: 6430408L10; A230052E19Rik; AI853486; B230332M13; BFIC3; BFIS3; BFNIS; C630029C19Rik; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; dmu; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSCI; HBSCII; HH1; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; Kiaa4197; LQT3; MED; mH1; mH2; microI; mKIAA4197; mnd-2; mnd2; Na(v)1.2; Na(V)1.4; Na(V)1.8; NAC1; NAC1A; NAC2; NAC3; NaCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NAV1.9; Nbna1; NCHVS; NE-NA; NENA; nmf2; nmf335; nmf58; NSS2; nur14; PFHB1; PN1; PN3; PN4; PN5; RII/RIIA; RNSCPIIR; SCAL; SCIII; SCN; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; SCN9A; ScpII; seal; SFNP; SkM1; SkM2; SMEI; SNS; SNS-2; SNS2; SSS1; VF1
UniProt ID: (Human) Q9Y5Y9, (Human) Q9UI33, (Human) P35498, (Human) Q99250, (Human) Q9NY46, (Human) P35499, (Human) Q14524, (Human) Q9UQD0, (Human) Q15858, (Rat) Q62968, (Mouse) Q6QIY3, (Rat) O88457, (Mouse) Q9R053, (Rat) P04774, (Mouse) A2APX8, (Rat) P04775, (Mouse) B1AWN6, (Mouse) A2ASI5, (Rat) P08104, (Mouse) Q9ER60, (Rat) P15390, (Rat) P15389, (Mouse) Q9JJV9, (Mouse) Q9WTU3, (Rat) O88420, (Mouse) Q62205, (Rat) O08562
Entrez Gene ID: (Human) 6336, (Human) 11280, (Human) 6323, (Human) 6326, (Human) 6328, (Human) 6329, (Human) 6331, (Human) 6334, (Human) 6335, (Rat) 29571, (Mouse) 20264, (Rat) 29701, (Mouse) 24046, (Rat) 81574, (Mouse) 20265, (Rat) 24766, (Mouse) 110876, (Mouse) 20269, (Rat) 497770, (Mouse) 110880, (Rat) 25722, (Rat) 25665, (Mouse) 20271, (Mouse) 20273, (Rat) 29710, (Mouse) 20274, (Rat) 78956
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.