Invitrogen
Manufacturer Code:MA112751
Catalog # 01-674-052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rabbit |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6H2 |
Immunogen | Fusion protein from the N-terminus of rabbit renal Na&plus/Ca2&plus exchanger. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Na/Ca exchanger; sodium/calcium exchanger; Sodium/Calcium Exchanger 1; solute carrier family 8 (sodium/calcium exchanger), member 1
Gene Aliases: NACA; NCX1
Entrez Gene ID: (Rabbit) 100328570
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.