Novus Biologicals
Manufacturer Code:NB3001270.02MG
Catalog # NB300127002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6H2 |
Immunogen | Fusion protein from the N-terminal region of rabbit renal NaCX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: Na(+)/Ca(2+)-exchange protein 1; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Sodium/calcium exchanger 1; solute carrier family 8 (sodium/calcium exchanger), member 1; Solute carrier family 8 member 1
Gene Aliases: CNC; NCX1; SLC8A1
UniProt ID: (Human) P32418
Entrez Gene ID: (Human) 6546
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.