Abnova
Manufacturer Code:PAB28989
Catalog # 89-130-499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protien corresponding rat Sod2. |
Conjugate | ATTO 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against Recombinant of Sod2.
For Research Use Only
Protein Aliases: mitochondrial superoxide dismutase 2; Superoxide dimutase 2, mitochondrial; Superoxide dismutase [Mn], mitochondrial
Gene Aliases: Sod2
UniProt ID: (Rat) P07895
Entrez Gene ID: (Rat) 24787
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.