R&D Systems
Manufacturer Code:MAB7795SP
Catalog # MAB7795SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 809116 |
Immunogen | <EM>E. coli-</EM>derived recombinant human Snapin Met1-Lys136 Accession # O95295 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 7; Biogenesis of lysosome-related organelles complex 1 subunit 7; BLOC-1 subunit 7; SNAP-25-binding protein; SNAP-25-binding protein SNAP25BP SNAPAP SNAPAPSNARE-associated protein Snapin SNAP-associated proteinSNARE associated protein snapin Synaptosomal-associated protein 25-binding protein; SNAP-associated protein; SNARE associated protein snapin; SNARE-associated protein Snapin; Synaptosomal-associated protein 25-binding protein
Gene Aliases: BLOC1S7; BLOS7; BORCS3; SNAP25BP; SNAPAP; SNAPIN
UniProt ID: (Human) O95295
Entrez Gene ID: (Human) 23557
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.