R&D Systems
Manufacturer Code:MAB10276100
Catalog # MAB10276100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1015418 |
Immunogen | Chinese Hamster Ovary cell line CHO-derived human Smoothened Met1-Tyr233 Accession # Q99835 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bnb CRJS FZD11 Gx Protein Gx SMO SMOH SMOHseven transmembrane helix receptor smoothened (Drosophila) homolog smoothened homolog smoothened homolog (Drosophila); frizzled family member 11; Protein Gx; seven transmembrane helix receptor; SMO; Smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: FZD11; Gx; SMO; SMOH
UniProt ID: (Human) Q99835
Entrez Gene ID: (Human) 6608
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.