Novus Biologicals
Manufacturer Code:NB10056688
Catalog # NB10056688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was raised against two peptides made from within amino acids 10 and 50 of human SMAD7. This sequence is 100% or highly conserved between human and multiple species. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CRCS3 FLJ16482 hSMAD7 MAD homolog 7 MAD homolog 8 MAD mothers against decapentaplegic homolog 7 (Drosophila) MADH7mothers against decapentaplegic homolog 7 MADH8 Mothers against decapentaplegic homolog 8 Mothers against decapentaplegic drosophila homolog of 7 Mothers against DPP homolog 7 Mothers against DPP homolog 8 SMAD 7 SMAD family member 7MAD (mothers against decapentaplegic Drosophila) homolog 7 SMAD mothers against DPP homolog 7 SMAD mothers against DPP homolog 7 (Drosophila) Smad7; hSMAD7; MAD (mothers against decapentaplegic, Drosophila) homolog 7; MAD homolog 7; MAD homolog 8; Mothers against decapentaplegic homolog 7; Mothers against decapentaplegic homolog 8; Mothers against decapentaplegic, drosophila, homolog of, 7; mothers against DPP homolog 8; SMAD 7; SMAD family member 7; SMAD, mothers against DPP homolog 7
Gene Aliases: CRCS3; MADH7; MADH8; SMAD7
UniProt ID: (Human) O15105
Entrez Gene ID: (Human) 4092
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.