Invitrogen
Manufacturer Code:PA517667
Catalog # PIPA517667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the residues surrounding Ile251 of human Smad5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JV5-1; MAD homolog 5; MAD homolog 5 MAD mothers against decapentaplegic homolog 5 OTTHUMP00000223331 SMA- and MAD-related protein 5 SMAD 5 SMAD mothers against DPP homolog 5 hSmad5 mothers against DPP homolog 5 mothers against decapentaplegic homolog 5 mothers against decapentaplegic drosophila homolog of 5; MAD, mothers against decapentaplegic homolog 5; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 5; SMA- and MAD-related protein 5; SMAD 5; SMAD family member 5; SMAD, mothers against DPP homolog 5
Gene Aliases: DWFC; JV5-1; MADH5; SMAD5
UniProt ID: (Human) Q99717, (Rat) Q9R1V3
Entrez Gene ID: (Human) 4090, (Rat) 59328
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.