R&D Systems
Manufacturer Code:AF2250
Catalog # AF2250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Smad5 Val155-Arg258 Accession # Q99717 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Dwfc hSmad5 JV5-1 JV5-1DKFZp781O1323 MAD homolog 5 MAD mothers against decapentaplegic homolog 5 (Drosophila) MADH5 MADH5mothers against decapentaplegic homolog 5 mothers against decapentaplegic homolog 5 mothers against decapentaplegic drosophila homolog of 5 Mothers against DPP homolog 5 SMA- and MAD-related protein 5 SMAD 5 SMAD family member 5DKFZp781C1895 SMAD mothers against DPP homolog 5 SMAD mothers against DPP homolog 5 (Drosophila) Smad5; JV5-1; MAD homolog 5; MAD, mothers against decapentaplegic homolog 5; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; SMA- and MAD-related protein 5; SMAD 5; SMAD family member 5; SMAD, mothers against DPP homolog 5
Gene Aliases: DWFC; JV5-1; MADH5; SMAD5
UniProt ID: (Human) Q99717
Entrez Gene ID: (Human) 4090
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.