Novus Biologicals
Manufacturer Code:NBP151646
Catalog # NBP151646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G1C6 |
Immunogen | Purified recombinant fragment of human SMAD4 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deleted in pancreatic carcinoma locus 4; Deletion target in pancreatic carcinoma 4; DPC4SMAD mothers against DPP homolog 4 (Drosophila) hSMAD4 JIP MAD homolog 4 MADH4mothers against decapentaplegic homolog 4 mothers against decapentaplegic homolog 4 mothers against decapentaplegic Drosophila homolog of 4 SMAD 4 SMAD family member 4MAD mothers against decapentaplegic homolog 4 (Drosophila) SMAD mothers against DPP homolog 4 Smad4; MAD homolog 4; Mothers against decapentaplegic homolog 4; mothers against decapentaplegic, Drosophila, homolog of, 4; SMAD 4; SMAD family member 4; SMAD, mothers against DPP homolog 4
Gene Aliases: DPC4; JIP; MADH4; MYHRS; SMAD4
UniProt ID: (Human) Q13485
Entrez Gene ID: (Human) 4089
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.