R&D Systems
Manufacturer Code:MAB4038
Catalog # MAB4038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 378611 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Smad3 Ser2-Ala230 Accession # P84022 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586N0721 DKFZp686J10186 hMAD-3 hSMAD3 HsT17436 JV15-2MGC60396 MAD homolog 3 mad protein homolog MAD mothers against decapentaplegic homolog 3 MAD mothers against decapentaplegic homolog 3 (Drosophila) mad3 MADH3mad homolog JV15-2 mothers against decapentaplegic homolog 3 Mothers against DPP homolog 3 SMA- and MAD-related protein 3 SMAD 3 SMAD family member 3HSPC193 SMAD mothers against DPP homolog 3 SMAD mothers against DPP homolog 3 (Drosophila) Smad3; hMAD-3; hSMAD3; JV15-2; MAD homolog 3; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; Mothers against decapentaplegic homolog 3; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD 3; SMAD family member 3; SMAD, mothers against DPP homolog 3
Gene Aliases: HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MADH3; SMAD3
UniProt ID: (Human) P84022
Entrez Gene ID: (Human) 4088
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.