Novus Biologicals
Manufacturer Code:NB10056462SS
Catalog # NB10056462T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amino acids 234-249 (DQQLNQSMDTGSPAEL) of human SMAD2 protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMAD-2; hMAD-2 hSMAD2 JV18 JV18-1SMAD mothers against DPP homolog 2 (Drosophila) MAD homolog 2 Mad protein homolog Mad mothers against decapentaplegic homolog 2 MADH2MGC34440 MADR2MGC22139 Mad-related protein 2 mother against DPP homolog 2 mothers against decapentaplegic homolog 2 Mothers against DPP homolog 2 Sma- and Mad-related protein 2 SMAD 2 SMAD family member 2MAD mothers against decapentaplegic homolog 2 (Drosophila) SMAD mothers against DPP homolog 2 Smad2; JV18-1; MAD homolog 2; Mad-related protein 2; mother against DPP homolog 2; Mothers against decapentaplegic homolog 2; Sma- and Mad-related protein 2; SMAD 2; SMAD family member 2; SMAD, mothers against DPP homolog 2
Gene Aliases: hMAD-2; hSMAD2; JV18; JV18-1; MADH2; MADR2; SMAD2
UniProt ID: (Human) Q15796
Entrez Gene ID: (Human) 4087
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.